Assay Details
Target Gene Details
Entrez Gene ID: | 11814 |
Gene Name: | apolipoprotein C-III |
Gene Aliases: |
apo-CIII, apoC-III |
Location: |
Chr.9:46233050-46235636 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apoc3 | NM_001289755.1 | NP_001276684.1 | ||
| NM_001289756.1 | NP_001276685.1 | |||
| NM_001289833.1 | NP_001276762.1 | |||
| NM_023114.4 | NP_075603.1 | |||
| AK002908.1 | BAB22448.1 | |||
| AK008260.1 | BAB25563.1 | |||
| BC021776.1 | AAH21776.1 | |||
| L04150.1 | ||||
| W61891.1 |
More Information
Set Membership: |
|

Genomic Map