Assay Details
Target Gene Details
Entrez Gene ID: | 73582 |
Gene Name: | calmodulin-lysine N-methyltransferase |
Gene Aliases: |
1700106N22Rik, AI480743, AV099781, CLNMT, CaM KMT |
Location: |
Chr.17:85090592-85458589 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Camkmt | NM_028576.2 | NP_082852.1 | ||
| AK007130.1 | BAB24867.1 | |||
| AK155246.1 | BAE33145.1 | |||
| AK172547.1 | ||||
| BC138272.1 | ||||
| BC138273.1 |
More Information
Set Membership: |
|

Genomic Map