Assay Details
Target Gene Details
Entrez Gene ID: | 14768 |
Gene Name: | LanC (bacterial lantibiotic synthetase component C)-like 1 |
Gene Aliases: |
AW124738, Gpr69, Gpr69a, p40 |
Location: |
Chr.1:67000517-67038872 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lancl1 | NM_001190984.1 | NP_001177913.1 | ||
| NM_001190985.1 | NP_001177914.1 | |||
| NM_021295.3 | NP_067270.1 | |||
| AJ294535.1 | CAC69241.1 | |||
| AK146120.1 | ||||
| AK163015.1 | 1 | 695 | ||
| AK182687.1 | ||||
| AK186065.1 | ||||
| AK210427.1 | ||||
| BC058560.1 | AAH58560.1 | |||
| Y11550.1 | CAA72314.1 | |||
| Y16518.1 | CAA76269.1 |
More Information
Set Membership: |
|

Genomic Map