Assay Details
Target Gene Details
Entrez Gene ID: | 101985 |
Gene Name: | U6 snRNA biogenesis 1 |
Gene Aliases: |
AA960436 |
Location: |
Chr.8:95332284-95347513 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usb1 | NM_133954.2 | NP_598715.2 | ||
| AK135527.1 | BAE22567.1 | |||
| AK160901.1 | BAE36080.1 | |||
| AK168683.1 | BAE40531.1 | |||
| BC016418.1 | AAH16418.1 |
More Information
Set Membership: |
|

Genomic Map