Assay Details
Target Gene Details
Entrez Gene ID: | 66460 |
Gene Name: | SYS1 Golgi-localized integral membrane protein homolog (S. cerevisiae) |
Gene Aliases: |
2610042O14Rik, AI266964, C87389 |
Location: |
Chr.2:164456981-164479637 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sys1 | AK032073.1 | |||
| AK032098.1 |
Target Gene Details
Entrez Gene ID: | 73603 |
Gene Name: | transformation related protein 53 target 5 |
Gene Aliases: |
1700126L10Rik |
Location: |
Chr.2:164467903-164478812 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trp53tg5 | NM_001271575.1 | NP_001258504.1 | ||
| AK007294.1 | BAB24938.1 |
More Information
Set Membership: |
|

Genomic Map