Assay Details
Target Gene Details
Entrez Gene ID: | 13401 |
Gene Name: | dystrophia myotonica-containing WD repeat motif |
Gene Aliases: |
DMR-N9, Dm9 |
Location: |
Chr.7:19076089-19082776 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dmwd | NM_010058.2 | NP_034188.2 | ||
| AK086899.1 | ||||
| BC089027.1 | AAH89027.1 | |||
| CF169673.1 | ||||
| S60312.1 | AAC60663.1 |
More Information
Set Membership: |
|

Genomic Map