Assay Details
Target Gene Details
Entrez Gene ID: | 271047 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade A, member 3B |
Gene Aliases: |
6A1, A030003A19Rik |
Location: |
Chr.12:104127531-104139546 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpina3b | NM_173024.3 | NP_766612.1 | ||
| AK037160.1 | BAC29726.1 | |||
| BC125418.1 | ||||
| BC145808.1 |
More Information
Set Membership: |
|

Genomic Map