Assay Details
Target Gene Details
Entrez Gene ID: | 18810 |
Gene Name: | plectin |
Gene Aliases: |
AA591047, AU042537, EBS1, PCN, PLTN, Plec1 |
Location: |
Chr.15:76170974-76231378 on Build GRCm38 |
Assay Gene Location: | Within Intron 17 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
