Assay Details
Target Gene Details
Entrez Gene ID: | 13803 |
Gene Name: | ectodermal-neural cortex 1 |
Gene Aliases: |
Nrpb, PIG10 |
Location: |
Chr.13:97241105-97253040 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Enc1 | NM_007930.4 | NP_031956.3 | ||
| AK008709.1 | ||||
| AK044906.1 | BAC32136.1 | |||
| AK049187.1 | BAC33597.1 | |||
| AK132173.1 | BAE21011.1 | |||
| BC049186.1 | AAH49186.1 | |||
| BC058098.1 | AAH58098.1 | |||
| U65079.1 | AAB64206.1 |
More Information
Set Membership: |
|

Genomic Map