Assay Details
Target Gene Details
Entrez Gene ID: | 107435 |
Gene Name: | histone aminotransferase 1 |
Gene Aliases: |
2410071B14Rik, AA536933, Hat-1, KAT1 |
Location: |
Chr.2:71389260-71441622 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hat1 | NM_026115.4 | NP_080391.2 | ||
| AK010715.1 | ||||
| AK014330.1 | ||||
| AK018270.1 | ||||
| AK041700.1 | BAC31038.1 | |||
| AK137648.1 | ||||
| AK169315.1 | BAE41070.1 | |||
| AK182724.1 | ||||
| BC055460.1 | AAH55460.1 |
More Information
Set Membership: |
|

Genomic Map