Assay Details
Target Gene Details
| Entrez Gene ID: | 16069 | 
| Gene Name: | immunoglobulin joining chain | 
| Gene Aliases: | 9530090F24Rik, AI323815, Igj, Jch | 
| Location: | Chr.5:88519809-88527897 on Build GRCm38 | 
| Assay Gene Location: | Within Intron 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Jchain | NM_152839.3 | NP_690052.2 | ||
| AB644392.1 | ||||
| AK078865.1 | BAC37428.1 | |||
| AK160344.1 | BAE35749.1 | |||
| BC006026.1 | AAH06026.1 | |||
| J00544.1 | AAA38673.1 | |||
| M90766.1 | AAA37918.1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  Validated | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top