Assay Details
Target Gene Details
Entrez Gene ID: | 16392 |
Gene Name: | ISL1 transcription factor, LIM/homeodomain |
Gene Aliases: |
- |
Location: |
Chr.13:116298257-116309721 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Isl1 | NM_021459.4 | NP_067434.3 | ||
| AB104633.1 | BAC57891.1 | |||
| AJ132765.1 | CAB38446.1 | |||
| AK090263.1 | BAC41153.1 | |||
| BC132263.1 | ||||
| BC132609.1 | ||||
| CF725499.1 |
More Information
Set Membership: |
|

Genomic Map