Assay Details
Target Gene Details
Entrez Gene ID: | 16667 |
Gene Name: | keratin 17 |
Gene Aliases: |
K17, Krt1-17 |
Location: |
Chr.11:100256217-100260989 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt17 | NM_010663.2 | NP_034793.1 | ||
| AB013608.1 | BAA34229.1 | |||
| BC132454.1 | ||||
| BC132456.1 |
More Information
Set Membership: |
|

Genomic Map