Assay Details
Target Gene Details
Entrez Gene ID: | 21334 |
Gene Name: | tachykinin 2 |
Gene Aliases: |
Tac3 |
Location: |
Chr.10:127724478-127731768 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tac2 | NM_001199971.1 | NP_001186900.1 | ||
| NM_009312.2 | NP_033338.2 | |||
| AF186116.1 | AAF01434.1 | |||
| BC051476.1 | AAH51476.1 | |||
| BQ175657.1 | ||||
| D14423.1 | BAA03316.1 |
More Information
Set Membership: |
|

Genomic Map