Assay Details
Target Gene Details
Entrez Gene ID: | 14841 |
Gene Name: | germ cell-specific gene 2 |
Gene Aliases: |
Haspin |
Location: |
Chr.11:73135484-73138294 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gsg2 | NM_010353.2 | 1 | 778 | NP_034483.1 |
| AK138750.1 | 1 | 779 | BAE23768.1 | |
| BC113148.1 | 1 | 772 | AAI13149.1 | |
| D87326.1 | 1 | 778 | BAA75494.1 |
Target Gene Details
Entrez Gene ID: | 16407 |
Gene Name: | integrin alpha E, epithelial-associated |
Gene Aliases: |
A530055J10, CD103, aM290, alpha-E1, alpha-M290 |
Location: |
Chr.11:73090595-73147449 on Build GRCm38 |
Assay Gene Location: | Within Intron 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itgae | NM_008399.2 | NP_032425.2 | ||
| AK154021.1 | BAE32321.1 | |||
| DQ386608.1 | ||||
| U12236.1 | AAC52142.1 |
More Information
Set Membership: |
|

Genomic Map