Assay Details
Target Gene Details
Entrez Gene ID: | 14582 |
Gene Name: | growth factor independent 1B |
Gene Aliases: |
Gfi-1B |
Location: |
Chr.2:28609450-28621982 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gfi1b | NM_001160406.1 | NP_001153878.1 | ||
| NM_008114.3 | NP_032140.1 | |||
| AF017275.1 | AAC40088.1 | |||
| AK157262.1 | BAE34016.1 | |||
| BC052654.1 | AAH52654.1 | |||
| BC132365.1 | ||||
| BC132367.1 | ||||
| BC145238.1 |
More Information
Set Membership: |
|

Genomic Map