Assay Details
Target Gene Details
Entrez Gene ID: | 66362 |
Gene Name: | exosome component 3 |
Gene Aliases: |
2310005D06Rik, AI593501, Rrp40 |
Location: |
Chr.4:45316629-45320603 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Exosc3 | NM_025513.3 | NP_079789.1 | ||
| AK006281.1 | BAB24501.1 | |||
| AK009165.1 | BAB26116.1 | |||
| BC023669.1 | AAH23669.1 | |||
| BC054085.1 | AAH54085.1 |
More Information
Set Membership: |
|

Genomic Map