Assay Details
Target Gene Details
Entrez Gene ID: | 23849 |
Gene Name: | Kruppel-like factor 6 |
Gene Aliases: |
AI448727, BCD1, C86813, CPBP, Copeb, FM2, FM6, Ierepo1, Ierepo3, R75280, Zf9 |
Location: |
Chr.13:5861489-5870393 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klf6 | NM_011803.2 | NP_035933.2 | ||
| AF072403.1 | AAC99478.1 | |||
| AK053584.1 | BAC35438.1 | |||
| AK144137.1 | BAE25722.1 | |||
| AK146615.1 | BAE27304.1 | |||
| AK147904.1 | BAE28217.1 | |||
| AK151769.1 | BAE30675.1 | |||
| AK159420.1 | BAE35068.1 | |||
| AY027436.1 | AAK11733.1 | |||
| BC020042.1 |
More Information
Set Membership: |
|

Genomic Map