Assay Details
Target Gene Details
Entrez Gene ID: | 16530 |
Gene Name: | potassium channel, subfamily K, member 7 |
Gene Aliases: |
2310014G03Rik, Kcnk6, Kcnk8, Knot, Mlk3 |
Location: |
Chr.19:5704476-5707405 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnk7 |

Genomic Map