Assay Details
Target Gene Details
Entrez Gene ID: | 240726 |
Gene Name: | solute carrier organic anion transporter family, member 5A1 |
Gene Aliases: |
4732454B15, A630033C23Rik, AA407151 |
Location: |
Chr.1:12866549-12992932 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slco5a1 | NM_172841.2 | NP_766429.2 | ||
| AK041736.1 | BAC31048.1 | |||
| AK048882.1 | BAC33482.1 | |||
| BC117943.1 | ||||
| BC117944.1 | ||||
| BQ031233.1 |
More Information
Set Membership: |
|

Genomic Map