Assay Details
| Gene Symbol: CPR76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_318993.4 | XP_318993.4 | 3 - 4 | 779 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CPR76| Gene Symbol: CPR76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_318993.4 | XP_318993.4 | 3 - 4 | 779 | 55 | |
Set Membership: |
|