Assay Details
Gene Symbol: MED29_ANOGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_321918.4 | XP_321918.4 | 2 - 3 | 623 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



