Assay Details
Gene Symbol: 1268694 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001687754.1 | XP_001687806.1 | 7 - 8 | 2249 | 75 | |
XM_307265.4 | XP_307265.4 | 6 - 7 | 2317 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
