Assay Details
| Gene Symbol: SPZ6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001688453.1 | XP_001688505.1 | 2 - 3 | 330 | 68 | |
| XM_314007.4 | XP_314007.3 | 2 - 3 | 331 | 68 | ||
More Information
Set Membership: |
|

SPZ6
Genomic Map