Assay Details
| Gene Symbol: CPR104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001688720.1 | XP_001688772.1 | 1 - 2 | 138 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CPR104| Gene Symbol: CPR104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001688720.1 | XP_001688772.1 | 1 - 2 | 138 | 62 | |
Set Membership: |
|