Assay Details
| Gene Symbol: OBP51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001688729.1 | XP_001688781.1 | 1 - 2 | 289 | 71 | |
| XM_316130.4 | XP_316130.4 | 2 - 3 | 244 | 71 | ||
More Information
Set Membership: |
|

OBP51
Genomic Map