Assay Details
| Gene Symbol: OBP17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001231182.1 | XP_001231183.1 | 1 - 2 | 249 | 65 | |
| XM_319537.4 | XP_319537.3 | 1 - 2 | 249 | 65 | ||
More Information
Set Membership: |
|

OBP17
Genomic Map