Assay Details
| Gene Symbol: ADT2_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003436049.1 | XP_003436097.1 | 3 - 4 | 622 | 52 | |
| XM_003436050.1 | XP_003436098.1 | 3 - 4 | 690 | 52 | ||
| XM_312601.5 | XP_312601.2 | 3 - 4 | 598 | 52 | ||
More Information
Set Membership: |
|

ADT2_ANOGA
Genomic Map