Assay Details
Gene Symbol: MED18_ANOGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_309758.5 | XP_309758.4 | 2 - 3 | 313 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
