Assay Details
| Gene Symbol: HPX11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_309791.5 | XP_309791.5 | 3 - 4 | 1411 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HPX11| Gene Symbol: HPX11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_309791.5 | XP_309791.5 | 3 - 4 | 1411 | 98 | |
Set Membership: |
|