Assay Details
| Gene Symbol: CTLGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001237814.2 | XP_001237815.1 | 2 - 3 | 376 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTLGA2| Gene Symbol: CTLGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001237814.2 | XP_001237815.1 | 2 - 3 | 376 | 65 | |
Set Membership: |
|