Assay Details
| Gene Symbol: OBP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001231180.2 | XP_001231181.2 | 1 | 319 | 65 | |
| XM_319533.2 | XP_319533.2 | 1 | 319 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OBP15| Gene Symbol: OBP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001231180.2 | XP_001231181.2 | 1 | 319 | 65 | |
| XM_319533.2 | XP_319533.2 | 1 | 319 | 65 | ||
Set Membership: |
|