Assay Details
| Gene Symbol: AT1G15370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101406.5 | NP_172989.1 | 3 - 4 | 513 | 68 | |
| GenBank mRNA | AF336926.1 | - | 3 - 4 | 244 | 68 | |
| AK118698.1 | - | 3 - 4 | 317 | 68 | ||
More Information
Set Membership: |
|

AT1G15370
Genomic Map