Assay Details
| Gene Symbol: CENP-C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101434.4 | NP_173018.2 | 10 - 11 | 2178 | 111 | |
| GenBank mRNA | AY693798.1 | - | 10 - 11 | 1980 | 111 | |
More Information
Set Membership: |
|

CENP-C
Genomic Map