Assay Details
| Gene Symbol: AVP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101437.5 | NP_173021.1 | 7 - 8 | 2293 | 63 | |
| GenBank mRNA | BT002481.1 | - | 7 - 8 | 2229 | 63 | |
| M81892.1 | - | 7 - 8 | 2259 | 63 | ||
More Information
Set Membership: |
|

AVP1
Genomic Map