Assay Details
Gene Symbol: SYP51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035977.1 | NP_001031054.1 | 2 - 3 | 402 | 72 | |
NM_001084077.1 | NP_001077546.1 | 2 - 3 | 425 | 72 | ||
NM_001332216.1 | NP_001323228.1 | 2 - 3 | 381 | 72 | ||
NM_101490.4 | NP_563994.1 | 2 - 3 | 428 | 72 | ||
GenBank mRNA | AF355755.1 | - | 2 - 3 | 338 | 72 | |
AK317530.1 | - | 2 - 3 | 403 | 72 | ||
AY074863.1 | - | 2 - 3 | 378 | 72 | ||
AY097382.1 | - | 1 - 2 | 212 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
