Assay Details
Gene Symbol: SYP51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035977.1 | NP_001031054.1 | 4 - 5 | 703 | 78 | |
NM_001084077.1 | NP_001077546.1 | 4 - 5 | 726 | 78 | ||
NM_001332216.1 | NP_001323228.1 | 4 - 5 | 682 | 78 | ||
NM_101490.4 | NP_563994.1 | 4 - 5 | 729 | 78 | ||
GenBank mRNA | AF355755.1 | - | 4 - 5 | 639 | 78 | |
AK317530.1 | - | 4 - 5 | 704 | 78 | ||
AY074863.1 | - | 4 - 5 | 679 | 78 | ||
AY085320.1 | - | 4 - 5 | 653 | 78 | ||
AY097382.1 | - | 3 - 4 | 513 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
