Assay Details
| Gene Symbol: RH36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101494.2 | NP_173078.1 | 5 - 6 | 1517 | 141 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RH36| Gene Symbol: RH36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101494.2 | NP_173078.1 | 5 - 6 | 1517 | 141 | |
Set Membership: |
|