Assay Details
Gene Symbol: RDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035979.1 | NP_001031056.1 | 6 - 7 | 750 | 75 | |
NM_001198083.1 | NP_001185012.1 | 7 - 8 | 823 | 75 | ||
NM_001332226.1 | NP_001322506.1 | 6 - 7 | 734 | 75 | ||
NM_001332227.1 | NP_001322507.1 | 6 - 7 | 744 | 75 | ||
NM_101511.6 | NP_563998.1 | 6 - 7 | 794 | 75 | ||
NM_179344.2 | NP_849675.1 | 6 - 7 | 734 | 75 | ||
GenBank mRNA | AB032865.1 | - | 6 - 7 | 656 | 75 | |
AJ010500.1 | - | 6 - 7 | 664 | 75 | ||
AK229282.1 | - | 6 - 7 | 753 | 75 | ||
AY084558.1 | - | 6 - 7 | 734 | 75 | ||
BT024876.1 | - | 5 - 6 | 591 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
