Assay Details
| Gene Symbol: HAC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198086.1 | NP_001185015.1 | 3 - 4 | 652 | 71 | |
| NM_001332245.1 | NP_001321740.1 | 3 - 4 | 883 | 71 | ||
| NM_001332246.1 | NP_001321737.1 | 3 - 4 | 1093 | 71 | ||
| NM_001332247.1 | NP_001321739.1 | 3 - 4 | 1093 | 71 | ||
| NM_001332248.1 | NP_001321738.1 | 4 - 5 | 937 | 71 | ||
| NM_001332249.1 | NP_001321742.1 | 3 - 4 | 698 | 71 | ||
| NM_001332250.1 | NP_001321741.1 | 3 - 4 | 649 | 71 | ||
| NM_001332251.1 | NP_001321736.1 | 3 - 4 | 929 | 71 | ||
| NM_001332252.1 | NP_001321735.1 | 3 - 4 | 910 | 71 | ||
| NM_101532.3 | NP_173115.1 | 3 - 4 | 598 | 71 | ||
More Information
Set Membership: |
|

HAC12
Genomic Map