Assay Details
Gene Symbol: EBS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101551.4 | NP_173134.2 | 8 - 9 | 1617 | 94 | |
GenBank mRNA | AY140061.1 | - | 8 - 9 | 1617 | 94 | |
BT010395.1 | - | 8 - 9 | 1487 | 94 | ||
KJ139003.1 | - | 8 - 9 | 1487 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
