Assay Details
| Gene Symbol: FUG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101583.4 | NP_173165.1 | 10 - 11 | 3117 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FUG1| Gene Symbol: FUG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101583.4 | NP_173165.1 | 10 - 11 | 3117 | 64 | |
Set Membership: |
|