Assay Details
| Gene Symbol: UBC34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084085.1 | NP_001077554.1 | 5 - 6 | 297 | 70 | |
| NM_001332287.1 | NP_001319027.1 | 5 - 6 | 572 | 70 | ||
| NM_001332288.1 | NP_001321628.1 | 5 - 6 | 737 | 70 | ||
| NM_001332289.1 | NP_001321631.1 | 5 - 6 | 608 | 70 | ||
| NM_001332290.1 | NP_001321629.1 | 5 - 6 | 1131 | 70 | ||
| NM_001332291.1 | NP_001321630.1 | 5 - 6 | 748 | 70 | ||
| NM_001332292.1 | NP_001321626.1 | 5 - 6 | 636 | 70 | ||
| NM_001332293.1 | NP_001321627.1 | 6 - 7 | 387 | 70 | ||
| NM_101590.4 | NP_173172.1 | 5 - 6 | 608 | 70 | ||
| GenBank mRNA | AK227344.1 | - | 5 - 6 | 316 | 70 | |
| AY084676.1 | - | 5 - 6 | 301 | 70 | ||
| BT003073.1 | - | 4 - 5 | 209 | 70 | ||
More Information
Set Membership: |
|

UBC34
Genomic Map