Assay Details
| Gene Symbol: UBC34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084085.1 | NP_001077554.1 | 8 - 9 | 435 | 110 | |
| NM_001332287.1 | NP_001319027.1 | 8 - 9 | 710 | 110 | ||
| NM_001332288.1 | NP_001321628.1 | 8 - 9 | 875 | 110 | ||
| NM_001332289.1 | NP_001321631.1 | 8 - 9 | 764 | 110 | ||
| NM_001332290.1 | NP_001321629.1 | 8 - 9 | 1269 | 110 | ||
| NM_001332291.1 | NP_001321630.1 | 8 - 9 | 886 | 110 | ||
| NM_001332292.1 | NP_001321626.1 | 8 - 9 | 774 | 110 | ||
| NM_001332293.1 | NP_001321627.1 | 9 - 10 | 525 | 110 | ||
| NM_101590.4 | NP_173172.1 | 8 - 9 | 746 | 110 | ||
| GenBank mRNA | AK227344.1 | - | 8 - 9 | 454 | 110 | |
| AY084676.1 | - | 8 - 9 | 439 | 110 | ||
| BT003073.1 | - | 7 - 8 | 347 | 110 | ||
More Information
Set Membership: |
|

UBC34
Genomic Map