Assay Details
Gene Symbol: EER4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101605.5 | NP_564023.1 | 2 - 3 | 1276 | 85 | |
NM_179349.1 | NP_849680.1 | 2 - 3 | 1113 | 85 | ||
GenBank mRNA | AY056144.1 | - | 2 - 3 | 1113 | 85 | |
AY091423.1 | - | 2 - 3 | 1005 | 85 | ||
AY463614.1 | - | 2 - 3 | 1005 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
