Assay Details
| Gene Symbol: HAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332312.1 | NP_001322988.1 | 4 - 5 | 1612 | 85 | |
| NM_101618.3 | NP_173199.2 | 3 - 4 | 1850 | 85 | ||
More Information
Set Membership: |
|

HAB2
Genomic Map