Assay Details
| Gene Symbol: CSTF77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101639.3 | NP_173218.2 | 3 - 4 | 575 | 69 | |
| GenBank mRNA | BT002320.1 | - | 3 - 4 | 229 | 69 | |
More Information
Set Membership: |
|

CSTF77
Genomic Map