Assay Details
| Gene Symbol: AT1G17860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101649.2 | NP_173228.1 | 1 | 566 | 77 | |
| GenBank mRNA | AF332416.1 | - | 1 | 524 | 77 | |
| AF370535.1 | - | 1 | 566 | 77 | ||
| AF410274.1 | - | 1 | 561 | 77 | ||
| AY081529.1 | - | 1 | 524 | 77 | ||
| AY097369.1 | - | 1 | 524 | 77 | ||
More Information
Set Membership: |
|

AT1G17860
Genomic Map