Assay Details
| Gene Symbol: PAPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101661.5 | NP_173240.2 | 3 - 4 | 584 | 104 | |
| GenBank mRNA | AK227446.1 | - | 3 - 4 | 584 | 104 | |
| AY323906.1 | - | 3 - 4 | 294 | 104 | ||
| BT005990.1 | - | 3 - 4 | 294 | 104 | ||
More Information
Set Membership: |
|

PAPS1
Genomic Map