Assay Details
| Gene Symbol: PAPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101661.5 | NP_173240.2 | 6 - 7 | 915 | 92 | |
| NM_202126.2 | NP_973855.1 | 4 - 5 | 950 | 92 | ||
| GenBank mRNA | AK227446.1 | - | 6 - 7 | 915 | 92 | |
| AY323906.1 | - | 6 - 7 | 625 | 92 | ||
| BT005990.1 | - | 6 - 7 | 625 | 92 | ||
More Information
Set Membership: |
|

PAPS1
Genomic Map